missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MP Biomedicals™ Human Serum: Hepatitis C (HCV)-Free
Hepatitis Associated Antigen (HBsAG)–free, HIV–free
Supplier: MP Biomedicals™ 092931149
Specifications
U.S. regulations | |
-20°C | |
Liquid | |
100 mL | |
0.1 μm filtration | |
≤26mg/dL | |
263 to 323mOsm/kg | |
5.5 to 7.5 g/dL |
Golden Yellow | |
Human | |
Human Serum | |
Sterile | |
Base matrix for hormone and steroid calibrators, controls, standards and linearity products | |
No | |
7 to 8 |