missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ Kanamycin Sulfate

Serve variety of purposes in cell culture, including selection, elimination of mycoplasma and microbial contamination, and prophylaxis

$61.39 - $158.89

Specifications

Content And Storage 15° to 30°C
Product Type Antibiotic
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN19453101
View Documents
MP Biomedicals™
0219453101
1 g
Each for $61.39
 
ICN19453105
View Documents
MP Biomedicals™
0219453105
5 g
Each for $158.89
 
Description

Description

All antibiotics are intended for research use only. They are not for therapeutic use in humans or animals.

Antibiotic effective against most gram positive and gram negative bacteria and most types of yeast.

Specifications

Specifications

15° to 30°C
Antibiotic
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'