missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ Lipase, USP Grade from Porcine Pancreas

For the resolution of chiral compounds and the transesterification production of biodiesel

Supplier:  MP Biomedicals™ 0210218980

 View more versions of this product

Catalog No. ICN10218980


Explore available promotions

Description

Description

Lipases are subclass of the esterases. It catalyzes the hydrolysis of emulsified esters of glycerol and long chain fatty acids.

  • Format: Cream powder
  • Partly soluble in water and insoluble in alcohol

Molecular biology

Specifications

Specifications

Cream
0°C
100 g
45000 to 50000
Powder
Used industrially for the resolution of chiral compounds and the transesterification production of biodiesel
Porcine pancreas
Lipase
Versene, Zn2+, Cu2+, Hg2+, iodine, PCMB
Videos
SDS
Documents

Documents

Promotions

Promotions