missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ Mycoplasma Removal Agent (MRA)

Mycoplasma Removal Agent (MRA) is a derivative of the quinoline family of antibiotics and eliminates mycoplasma contamination in cell cultures by inhibiting mycoplasma DNA gyrase.

Supplier:  MP Biomedicals™ 093050044

Catalog No. ICN3050044


Explore available promotions

Description

Description

  • Derivative of the quinoline family of antibiotics and eliminates mycoplasma infection by inhibiting mycoplasma DNA gyrase
  • Supplied as a ready-to-use solution
  • Cited by the European Collection of Animal Cell Cultures (Porton Down, U.K.) as being superior to all other current products for the eradication of mycoplasma
  • Cell toxicity is low following treatment at the recommended concentration of 0.5 μg/mL
  • Eliminates multiple mycoplasma species within one week using the lowest dosage
  • Prevents recontamination of the culture at 0.1 μg/mL
  • Maintains cell viability
  • Compatible with most mammalian cell lines
  • Sterility and low cytotoxicity
Specifications

Specifications

Store at Room Temperature (15 to 30°C). Protect from light.
Clear, light yellow solution
Sterile
5 mL
50 µg/mL
Mycoplasma Removal
Mycoplasma elimination
Videos
SDS
Documents

Documents

Promotions

Promotions