missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ Proteinase K

Suitable for both protein and nucleic acid isolation

Supplier:  MP Biomedicals™ 0219350480

 View more versions of this product

Catalog No. ICN19350480


Explore available promotions

Description

Description

Proteinase K, [39450-01-6]
Proteolytic Activity: >30mAnson Units/mg
Unit Definition: One Anson unit is the amount of enzyme which liberates 1 μmol of Folin-positive amino acids per minute at pH 7.5 and 35°C, using hemoglobin as substrate

  • From Tritirachium album
  • Chromatographically purified
  • Exhibits proteolytic activity on proteins, peptides, glycoproteins, amides and esters.
  • Active with nitroanilides of amino acids with protected amino groups, excluding arginine.
  • Useful in the isolation of DNA and RNA, in the analysis of membrane structures and protein structure.

Specifications

Specifications

Protein Structure Analysis
Proteinase K
Chromatography
Tritirachium album Limber
Lyophilized
∽30
7.5 to 12
100 mg
4°C
Videos
SDS
Documents

Documents

Promotions

Promotions