missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ QuickFlex™ Adapter for 96 x 2 mL Tube Holder on FastPrep 96
QuickFlex™ adapter for 96 x 2 mL tube holder on FastPrep 96 is suited for DNA and RNA isolation, enzyme isolation and protein production, natural products isolation, food preparation for quality analysis, biopharma manufacturing, and forensic applications.
Supplier: MP Biomedicals™ 116010570
Description
- Applications: QuickFlex™ 96 x 2 mL Adapter for FastPrep-96™ is a large sample volume adapter is ideally suited for DNA and RNA isolation, enzyme isolation and protein production, natural products isolation, food preparation for quality analysis, biopharma manufacturing, and forensic applications. Additionally, frozen samples in Lysing Matrix tubes loaded in the adapter and stored at -20°C or -80°C are ready to be immediately processed with minimal hands-on manipulation, preventing degradation of cellular components by endogenous enzymes.
- Product Description: High-throughput adapter for the FastPrep-96™ that holds 96 x 2 mL lysing matrix tubes. The ergonomic design of the adapter ensures an easy loading of lysing matrix tubes that remain securely sealed during the homogenization. It stand stable on the bench top and is commonly used as a tube rack. Ideally suited for high-throughput applications, up to 96 samples can be homogenized simultaneously. Additionally, frozen samples in Lysing Matrix tubes loaded in the adapter and stored at -20°C or -80°C are ready to be immediately processed with minimal hands-on manipulation, preventing degradation of cellular components by endogenous enzymes.
- Capacity: 96 tubes
- Sample Volume: Up to 20 g
Specifications
| FastPrep-96 Adapters | |
| Room Temperature | |
| Extractions of nucleic acids, proteins, metabolites and other small molecules from samples |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.