missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ Ribonuclease A, From Bovine Pancreas

Aggregates can be converted to monomers.

Supplier:  MP Biomedicals™ 0210107680

 View more versions of this product

Catalog No. ICN10107680


Explore available promotions

Description

Description

Ribonuclease A, [9001-99-4]

  • RNase A
  • From Bovine Pancreas
  • Prepared from aggregate-free RNase
  • Lyophilized
  • Activity: 50 Kunitz units/mg.

Specifications

Specifications

≥50 kU/mg
>70%
Used to remove RNA from DNA plasmid preparations and Protein samples
Bovine pancreas
Ribonuclease A
Heavy Metal ions and is competitively inhibited by DNA
White
7 to 7.5
0°C
100 mg
13700
Lyophilized Powder
Videos
SDS
Documents

Documents

Promotions

Promotions