MP Biomedicals™ Roll & Grow™ Plating Beads
Safely and easily spread yeast and bacterial cells
Supplier:  MP Biomedicals™ 115000550
Catalog No. MP115000550
Explore available promotions
Description
Description
Roll & Grow™ Plating Beads are used to spread 25-400μL of bacterial or yeast cells across the surface of an agar plate, providing a safer, faster alternative to the traditional method of plating with a glass rod sterilized with ethanol and flame. Sterile Roll & Grow™ Plating Beads are provided in two skirted 50mL conical tubes. Between six and seven 4mm glass beads are used per plate for a total of approximately 160 standard 100mm plates.
  • Spread cells in just seconds per plate
  • Eliminate hazards of flame and ethanol
  • No need for fragile glass “hockey sticks”
  • Consistent, even colony distribution
  • Sterile and easy to use
Cell cycle; Cell proliferation; Cell metabolism; Cell viability; Cytotoxicity; Protein Trafficking; Bacteria cell culture; Cell preservation; Cell counting; Viability; Cryopreservation; Insect cell culture; Mammalian cell culture; Plant cell growth; Primary cell culture; Stem cell culture; Yeast cell culture
Specifications
Specifications
2 Tubes
Roll and Grow Plating Beads
Skirted 50mL conical tubes
15° to 30°C
2 Tubes
Glass
Cell cycle, cell proliferation, cell metabolism, cell viability, cytotoxicity, protein trafficking, bacteria cell culture, cell preservation, cell counting, viability, cryopreservation, insect cell culture, mammalian cell culture, plant cell growth
4mm
Sterile