missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ Trypsin from Beef Pancreas

2x Crystallized, ≥180 TAMEu/mg , White Powder. Commonly used in biological research.

$102.18 - $102.18

Specifications

Color White
Product Type Trypsin
Molecular Weight (g/mol) 23000 or 22000
View More Specs

Products 1
Catalog Number Mfr. No. Concentration Quantity Source Price Quantity  
Catalog Number Mfr. No. Concentration Quantity Source Price Quantity  
ICN1011791
View Documents
MP Biomedicals™
021011791
2X 100 mg Human, Porcine or Bovine pancreas
Each for $102.18
 
Description

Description

Specifications

Specifications

White
Trypsin
23000 or 22000
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'