MP Biomedicals™ Yeast Hydrolysate Enzymatic
Autolyzed Brewers yeast
Supplier:  MP Biomedicals™ 0210330425
 View more versions of this product
Catalog No. ICN10330425
Explore available promotions
Description
Description
Yeast Hydrolysate Enzymatic is a concentrated source of soluble protein components, carbohydrates, and B Complex vitamins natural to yeast.
Specifications
Specifications
15-30°C
Tan powder
25 lb.
Brewers Yeast