missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MSA™ Comfo™ and Comfo Classic™ Half-Mask Respirators
Available in Hycar™ rubber or silicone, Comfo™ series half-mask respirators feature a unique face seal design for a comfortable fit.
$106.19 - $106.19
Specifications
Certifications (NIOSH) | NIOSH 42 CFR Part 84 |
---|---|
Color | Black |
Product Type | Half-Mask Respirator |
For Use With (Application) | Utilities, oil and gas, construction, general industry, welding |
For Use With (Equipment) | Comfo™ Series cartridges and filters |
Catalog Number | Mfr. No. | Size | Facepiece Material | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Facepiece Material | Price | Quantity | ||||
189992588A
|
MSA™
808075 |
Small | Hycar™ |
Each for $106.19
|
|
||||
189992588B
|
MSA™
808074 |
Medium | Hycar™ |
Each for $106.19
|
|
||||
189992588C
|
MSA™
808076 |
Large | Hycar™ |
Each for $106.19
|
|
||||
189991511
|
MSA™
808072 |
Small | Silicone |
Each for $106.19
|
|
||||
189991510
|
MSA™
808071 |
Medium | Silicone |
Each for $106.19
|
|
||||
189991509
|
MSA™
808073 |
Large | Silicone |
Each for $106.19
|
|
||||
Description
- Protection from exposure to vapors, gases, and particulates
- SoftFeel™ facepiece material increases softness
- Choose from Hycar™ rubber or silicone
- Face seal design provides tight, comfortable fit
- Ideal markets include utilities, oil & gas, construction and general industry
Certifications
NIOSH approved under 42CFR84 for use against a variety of respiratory hazards, when used as designed with appropriate cartridges and filters.
Specifications
NIOSH 42 CFR Part 84 | |
Half-Mask Respirator | |
Comfo™ Series cartridges and filters |
Black | |
Utilities, oil and gas, construction, general industry, welding |