missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MSA™ Confidence Plus™ Germicidal Cleaner

Germicidally effective cleaning, sanitizing, and disinfecting solution for use with respiratory equipment.

Supplier:  MSA™ 10032737

Catalog No. 18-616T


Explore available promotions

Description

Description

  • Cost-effective, one-step disinfectant, cleaner, sanitizer
  • Built-in measuring cup
  • Ideal for faceshields, goggles, gas masks, SCBAs
  • Active ingredients: N-alkyl dimethyl benzyl ammonium chloride: 5%
To use:
  • Mix 1 oz. (30mL) of Confidence Plus with 1 gallon (3.8L) of warm water
  • Solution needs ten minutes of contact time for use as a disinfectant
  • Rinse in clean water and air dry after use

Specifications

Specifications

Germicidal Cleaner
32 oz.
Faceshields, goggles, gas masks, SCBAs
10009971
0.9 L
EPA
Videos
SDS
Documents

Documents

Promotions

Promotions