missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MSA™ Ultra Elite™ Twin Cartridge Full-Facepiece Respirators
When used with appropriate filter cartridges, the NIOSH-approved full facepiece respirator combination provides an ideal defense against numerous hazards including particulates, dust, projectiles, and splashing liquids in toxic atmospheres not deemed Immediately Dangerous to Life or Health (non-IDLH).
Supplier: MSA™ 10016755
Includes: Twin cartridge adapter
Requires : Comfo series aerosol filters and chemical cartridges
Description
- Ergonomic design makes this a comfortable unit
- Five-point rubber head harness makes it easy to put it on or take it off
- Distortion-free coated lens provides a wide field of vision
- Unique airflow prevents lens misting
- Face blank has a single-sealing flange made of soft-feel silicone
- Speech diaphragm makes communication easy to discern
- Suits a wide range of industrial applications including asbestos abatement, paper and pulp manufacturing and spray painting
Order Info
Additional accessories such as lens cover (Cat. No. 19-815-684) and speech diaphragm (Cat. No. 18-999-3861) available separately.
Specifications
Class 3 | |
Silicone | |
Small | |
Filter cartridge | |
10016755 |
NIOSH 42 CFR Part 84 | |
Full Facepiece Respirator with Adapter | |
Utilities, mining, oil and gas, fire service, construction, search and rescue, welding | |
Twin cartridge adapter | |
5-point |