missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MilliporeSigma™ MTT, Calbiochem™,

Membrane-permeable yellow dye that is reduced by mitochondrial reductases in living cells to form the dark blue product, MTT-formazan

Supplier:  MilliporeSigma™ 4759891GM

 View more versions of this product

Catalog No. 4759891GM


Explore available promotions

Description

Description

Specifications

Specifications

MTT
195°C
C18H16BrN5S
1 g
AZKSAVLVSZKNRD-UHFFFAOYSA-M
2-(4,5-dimethyl-1,3-thiazol-2-yl)-3,5-diphenyl-3H-1,2λ⁵,3,4-tetrazol-2-ylium bromide
64965
414.3
Solid
298-93-1
White
MFCD00011964,MFCD00066662
thiazolyl blue, thiazolyl blue tetrazolium bromide, mmt tetrazolium, methylthiazoletetrazolium, mtt, thiazolyl blue monotetrazolium, thiazole blue, 3-4,5-dimethylthiazol-2-yl-2,5-diphenyltetrazolium bromide, mtt van, unii-euy85h477i
[Br-].CC1=C(C)N=C(S1)[N+]1=NC(=NN1C1=CC=CC=C1)C1=CC=CC=C1
414.33
CHEBI:53233
≥98%

Chemical Identifiers

298-93-1
414.33
AZKSAVLVSZKNRD-UHFFFAOYSA-M
64965
2-(4,5-dimethyl-1,3-thiazol-2-yl)-3,5-diphenyl-3H-1,2λ⁵,3,4-tetrazol-2-ylium bromide
C18H16BrN5S
MFCD00011964,MFCD00066662
thiazolyl blue, thiazolyl blue tetrazolium bromide, mmt tetrazolium, methylthiazoletetrazolium, mtt, thiazolyl blue monotetrazolium, thiazole blue, 3-4,5-dimethylthiazol-2-yl-2,5-diphenyltetrazolium bromide, mtt van, unii-euy85h477i
CHEBI:53233
[Br-].CC1=C(C)N=C(S1)[N+]1=NC(=NN1C1=CC=CC=C1)C1=CC=CC=C1
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 206-069-5

RTECSNumber : XF8060000

Recommended Storage : 15° to 30°C (59° to 86°F); Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above 20°C (68°F).

SDS
Documents

Documents

Promotions

Promotions