missing translation for 'onlineSavingsMsg'
Learn More

Mucochloric acid, 99% (dry wt.), water <4.0%, Thermo Scientific Chemicals

$192.42 - $192.42

Chemical Identifiers

CAS 87-56-9
Molecular Formula C4H2Cl2O3
Molecular Weight (g/mol) 168.96
MDL Number MFCD00006966,MFCD00006966
InChI Key LUMLZKVIXLWTCI-IHWYPQMZSA-N
Synonym mucochloric acid, 2,3-dichloromaleic aldehyde acid, kyselina mukochlorova, 2z-2,3-dichloro-4-oxobut-2-enoic acid, 2-butenoic acid, 2,3-dichloro-4-oxo-, 2z, aldehydodichloromaleic acid, dichloromalealdehydic acid, unii-5i5877jhiw, kyselina mukochlorova czech, ccris 6597
PubChem CID 2771871
SMILES OC(=O)C(\Cl)=C(\Cl)C=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2452336
View Documents
Thermo Scientific Chemicals
B2452336
500 g
Each for $192.42
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

87-56-9
168.96
LUMLZKVIXLWTCI-IHWYPQMZSA-N
2771871
C4H2Cl2O3
MFCD00006966,MFCD00006966
mucochloric acid, 2,3-dichloromaleic aldehyde acid, kyselina mukochlorova, 2z-2,3-dichloro-4-oxobut-2-enoic acid, 2-butenoic acid, 2,3-dichloro-4-oxo-, 2z, aldehydodichloromaleic acid, dichloromalealdehydic acid, unii-5i5877jhiw, kyselina mukochlorova czech, ccris 6597
OC(=O)C(\Cl)=C(\Cl)C=O

Specifications

87-56-9
1.85
C4H2Cl2O3
500 g
1705641
mucochloric acid, 2,3-dichloromaleic aldehyde acid, kyselina mukochlorova, 2z-2,3-dichloro-4-oxobut-2-enoic acid, 2-butenoic acid, 2,3-dichloro-4-oxo-, 2z, aldehydodichloromaleic acid, dichloromalealdehydic acid, unii-5i5877jhiw, kyselina mukochlorova czech, ccris 6597
OC(=O)C(\Cl)=C(\Cl)C=O
168.96
168.96
Mucochloric acid
125°C to 128°C (dried)
186°C (367°F)
MFCD00006966,MFCD00006966
UN2923
14,6297
LUMLZKVIXLWTCI-IHWYPQMZSA-N
(Z)-2,3-dichloro-4-oxobut-2-enoic acid
2771871
99%
(dry wt.), water <4.0%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H314-H318-H341
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of causing genetic defects.

P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c

H301-H314-H317-H335-H340

DOTInformation : Hazard Class: 8; Packaging Group: II

EINECSNumber : 201-752-4

RTECSNumber : AS7700000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only