missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

OHAUS™ Multi-Position Stirrer

Use the Multi-Position Stirrer for synchronized and uniform mixing.

Supplier:  OHAUS™ 30573794

 View more versions of this product

Catalog No. 01918308


Explore available promotions

Description

Description

  • Applications: Dissolution Studies, Titrations, Media/Reagent Preparations, Synchronized Stirring of multiple samples
  • Display: Independent LED displays for speed and time (on digital models); Knob with 1-10 dial markings (on non-digital models)
  • Operation: N/A
  • Communication: N/A
  • Construction: Cast aluminum housing, glass filled nylon top plate. Includes stir bars to accommodate each stirring position
  • Design Features: Speed ramping feature slowly increases motor speed for enhanced coupling and improved stirring. Microprocessor-controlled for precise speed regulation. Timer counts up or when programmed to a set time will turn off stirring and sound alarm when timer reaches zero.
Specifications

Specifications

60 to 1400 rpm
9
Glass-filled Nylon
250 mL
120 V,115 V
0.1 g
AC adapter
ABS Plastic, Stainless Steel, Glass
4100 g
970 mm
17 in.
432 mm
12.5 in.
2 Years
11 in.
11 in.
1.3 A
18 cm
Analog
14 lb.
115 V 50/60 Hz
3 Button keypad
UL, cUL
RS-232
LCD
75 W
Milligram, gram, ounce, carat, pennyweight, troy o
±0.1 g
STMP9AL
4 in.
3 sec.
50°F to 104°F
5°C to 40°C, 80% RH, Non-condensing
Precision Balance
318 mm
279 mm
279 mm
50/60 Hz
12.6 in.
32 cm
6.3 kg
1 sec. to 160 hr.
Videos
SDS
Documents

Documents

Promotions

Promotions