Beckman Coulter Multisizer™ 4e Smart Technology High Resolution Aperture Tubes
Smart Technology High Resolution Aperture Tubes are designed specifically for the Multisizer 4e Analyzer. For greater discrimination of particle size, high resolution tubes have an aperture thickness 120% greater than normal apertures which extends the time it takes for the particle to pass through.
Supplier:  Beckman Coulter C93981
 View more versions of this product
Catalog No. 15655143
Explore available promotions
Description
Description
  • Extended measurement range (2% to 80% of aperture diameter) leads to less frequent aperture changes for more analysis options
  • A wide range of apertures is available with diameters ranging from 10 to 2,000 μm
  • Specifically designed for the Multisizer 4e or Multisizer 4 Coulter Counters
  • 100 μm ST Aperture Tube is standard with the Multisizer 4e Coulter Counter
  • Compatible with ST Beakers and Accuvette STs
  • Convenient tubes ensure accurate, and consistent results
  • For greater discrimination of particle size, high resolution tubes have an aperture thickness 120% greater than normal apertures which extends the time the particle takes to pass through
  • Size and serial number information is included on a bar code on the tube for automatic data entry via instrument bar code reader
Specifications
Specifications
High Resolution Aperature Tube
2.8 μm to 112 μm
Multisizer™ 4e Coulter Counters
Glass
140 μm