missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MP Biomedicals™ Murashige and Skoog Media
All Murashige and Skoog plant cell culture Media are manufactured from high quality chemicals and reagents.
Supplier: MP Biomedicals™ 092610022
Description
- Each formulation is prepared according to the original specification, except where noted
- Finished lots of media are plant cell tested
Specifications
4°C | |
Murashige and Skoog Medium | |
Solution | |
1 x 10 L |
Powder | |
No Sucrose, No Agar, No IAA, No Kinetin | |
4.80 to 5.65 |