missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Murexide, Thermo Scientific Chemicals

$101.60 - $1,219.26

Chemical Identifiers

CAS 3051-09-0
Molecular Formula C8H3N5O6-2
Molecular Weight (g/mol) 265.141
MDL Number MFCD00012777
InChI Key MUMMBXDKKJEASE-UHFFFAOYSA-L
Synonym murexide, ammonium purpurate acid, 6-oxo-5-2,4,6-trioxo-1,3-diazinan-5-ylidene amino-1h-pyrimidine-2,4-diolate, 5-hexahydro-2,4,6-trioxo-5-pyrimidinyl imino-2,4,6 1h,3h,5h-pyrimidinetrione ammonium salt, 6-oxidanylidene-5-2,4,6-tris oxidanylidene-1,3-diazinan-5-ylidene amino-1h-pyrimidine-2,4-diolate
PubChem CID 51381221
IUPAC Name 6-oxo-5-[(2,4,6-trioxo-1,3-diazinan-5-ylidene)amino]-1H-pyrimidine-2,4-diolate
SMILES C1(=C(N=C(NC1=O)[O-])[O-])N=C2C(=O)NC(=O)NC2=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1754009
View Documents
Thermo Scientific Chemicals
A1754009
10 g N/A
 
AAA1754018
View Documents
Thermo Scientific Chemicals
A1754018
50 g N/A
 
AAA1754030
View Documents
Thermo Scientific Chemicals
A1754030
250 g N/A
 
Description

Description

Murexide is used as a complexometric indicator in analytical chemistry for complexometric titrations of calcium, copper nickel and cobalt. It is an inhibitor of dihydrofolate reductase. Further, it acts as an enhancer of sonochemical destruction of chlorinated hydrocarbon pollutants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Murexide is used as a complexometric indicator in analytical chemistry for complexometric titrations of calcium, copper nickel and cobalt. It is an inhibitor of dihydrofolate reductase. Further, it acts as an enhancer of sonochemical destruction of chlorinated hydrocarbon pollutants.

Solubility
Soluble in water and 1M sodium hydroxide solution. Insoluble in alcohol.

Notes
Incompatible with strong oxidizing agents.                  
Specifications

Chemical Identifiers

3051-09-0
265.141
MUMMBXDKKJEASE-UHFFFAOYSA-L
51381221
C1(=C(N=C(NC1=O)[O-])[O-])N=C2C(=O)NC(=O)NC2=O
C8H3N5O6-2
MFCD00012777
murexide, ammonium purpurate acid, 6-oxo-5-2,4,6-trioxo-1,3-diazinan-5-ylidene amino-1h-pyrimidine-2,4-diolate, 5-hexahydro-2,4,6-trioxo-5-pyrimidinyl imino-2,4,6 1h,3h,5h-pyrimidinetrione ammonium salt, 6-oxidanylidene-5-2,4,6-tris oxidanylidene-1,3-diazinan-5-ylidene amino-1h-pyrimidine-2,4-diolate
6-oxo-5-[(2,4,6-trioxo-1,3-diazinan-5-ylidene)amino]-1H-pyrimidine-2,4-diolate

Specifications

>300°C
C8H3N5O6-2
3582175
murexide, ammonium purpurate acid, 6-oxo-5-2,4,6-trioxo-1,3-diazinan-5-ylidene amino-1h-pyrimidine-2,4-diolate, 5-hexahydro-2,4,6-trioxo-5-pyrimidinyl imino-2,4,6 1h,3h,5h-pyrimidinetrione ammonium salt, 6-oxidanylidene-5-2,4,6-tris oxidanylidene-1,3-diazinan-5-ylidene amino-1h-pyrimidine-2,4-diolate
MUMMBXDKKJEASE-UHFFFAOYSA-L
6-oxo-5-[(2,4,6-trioxo-1,3-diazinan-5-ylidene)amino]-1H-pyrimidine-2,4-diolate
51381221
10 g
3051-09-0
MFCD00012777
14,6306
Soluble in water and 1M sodium hydroxide solution. Insoluble in alcohol.
C1(=C(N=C(NC1=O)[O-])[O-])N=C2C(=O)NC(=O)NC2=O
265.141
284.19
Murexide
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 221-266-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only