Learn More
Thermo Scientific Chemicals Mycophenolic acid, 98%
CAS: 24280-93-1 | C17H20O6 | 320.34 g/mol
$214.64 - $214.64
Identifiants chimiques
| CAS | 24280-93-1 |
|---|---|
| Molecular Formula | C17H20O6 |
| Molecular Weight (g/mol) | 320.34 |
| InChI Key | HPNSFSBZBAHARI-RUDMXATFSA-N |
| Synonym | mycophenolic acid, mycophenolate, melbex, myfortic, mycophenolsaeure, acido micofenolico, micofenolico acido, acide mycophenolique, acidum mycophenolicum, 6-4-hydroxy-6-methoxy-7-methyl-3-oxo-5-phthalanyl-4-methyl-4-hexenoic acid |
| PubChem CID | 446541 |
| ChEBI | CHEBI:168396 |
| IUPAC Name | (E)-6-(4-hydroxy-6-methoxy-7-methyl-3-oxo-1H-2-benzofuran-5-yl)-4-methylhex-4-enoic acid |
| SMILES | CC1=C(C(=C(C2=C1COC2=O)O)CC=C(C)CCC(=O)O)OC |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|
AC459380050
|
Thermo Scientific Chemicals
459380050 |
5 g |
chaque for $214.64
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identifiants chimiques
| 24280-93-1 | |
| 320.34 | |
| mycophenolic acid, mycophenolate, melbex, myfortic, mycophenolsaeure, acido micofenolico, micofenolico acido, acide mycophenolique, acidum mycophenolicum, 6-4-hydroxy-6-methoxy-7-methyl-3-oxo-5-phthalanyl-4-methyl-4-hexenoic acid | |
| CHEBI:168396 | |
| CC1=C(C(=C(C2=C1COC2=O)O)CC=C(C)CCC(=O)O)OC |
| C17H20O6 | |
| HPNSFSBZBAHARI-RUDMXATFSA-N | |
| 446541 | |
| (E)-6-(4-hydroxy-6-methoxy-7-methyl-3-oxo-1H-2-benzofuran-5-yl)-4-methylhex-4-enoic acid |
Spécifications
| 24280-93-1 | |
| Authentic | |
| C17H20O6 | |
| mycophenolic acid, mycophenolate, melbex, myfortic, mycophenolsaeure, acido micofenolico, micofenolico acido, acide mycophenolique, acidum mycophenolicum, 6-4-hydroxy-6-methoxy-7-methyl-3-oxo-5-phthalanyl-4-methyl-4-hexenoic acid | |
| CC1=C(C(=C(C2=C1COC2=O)O)CC=C(C)CCC(=O)O)OC | |
| 320.34 | |
| CHEBI:168396 | |
| 98% |
| 0.5% max. | |
| 97.5% min. (HPLC) | |
| 5 g | |
| HPNSFSBZBAHARI-RUDMXATFSA-N | |
| (E)-6-(4-hydroxy-6-methoxy-7-methyl-3-oxo-1H-2-benzofuran-5-yl)-4-methylhex-4-enoic acid | |
| 446541 | |
| 320.34 | |
| Mycophenolic acid |
Sécurité et manipulation
GHS H Statement
Harmful if swallowed.
May damage the unborn child.
Suspected of causing genetic defects.
Causes damage to organs through prolonged or repeated exposure if swallowed.
Very toxic to aquatic life with long last
GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Use personal protective equipment as required.
IF exposed or concerned: Get m
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 246-119-3
RUO – Research Use Only