missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Novex™ MYR PKC Zeta Pseudosubstrate DFS Item

MYR PKC Zeta Pseudosubstrate

Supplier:  Novex™ LS77749

Catalog No. 77749


Explore available promotions

Description

Description

Selective protein kinase C (PKC) zeta inhibitor.

Format/Formulation: Lyophilized trifluoroacetate salt, >97% purity by HPLC.

Molecular Weight: 1928
Specifications

Specifications

Store in freezer (-5 to -30°C).
1 mg
Peptide
1928 Da
Dry Ice
Lyophilized
5590
Kinase Inhibitor (PKC Inhibitor), Kinase Inhibitor
HPLC
PKCζ
Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.