Learn More
Myrcene, 90%, tech., stabilized, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 128080025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Myrcene | |
Colorless to Yellow | |
165.0°C | |
Authentic | |
Glass bottle | |
1.4660 to 1.4720 | |
2.5 L | |
0.795 | |
myrcene, beta-myrcene, 7-methyl-3-methyleneocta-1,6-diene, 7-methyl-3-methylene-1,6-octadiene, 1,6-octadiene, 7-methyl-3-methylene, beta-geraniolene, .beta.-myrcene, 2-methyl-6-methylene-2,7-octadiene, 3-methylene-7-methyl-1,6-octadiene, myrcene natural | |
UAHWPYUMFXYFJY-UHFFFAOYSA-N | |
7-methyl-3-methylideneocta-1,6-diene | |
Stabilized with 0.01% alpha-Tocopherol | |
CHEBI:17221 | |
90% |
123-35-3,127-91-3,138-86-3 | |
0.7950g/mL | |
44°C | |
88% min. (GC) | |
C10H16 | |
H2C=CHC(=CH2)CH2CH2CH=C(CH3)2 | |
01,264 | |
15,6416 | |
Solubility in water: practically insoluble. Other solubilities: soluble in alcohol, chloroform, ether and, glacial acetic acid | |
CC(=CCCC(=C)C=C)C | |
136.24 | |
31253 | |
136.24 | |
Liquid |
Chemical Identifiers
123-35-3 | |
136.24 | |
myrcene, beta-myrcene, 7-methyl-3-methyleneocta-1,6-diene, 7-methyl-3-methylene-1,6-octadiene, 1,6-octadiene, 7-methyl-3-methylene, beta-geraniolene, .beta.-myrcene, 2-methyl-6-methylene-2,7-octadiene, 3-methylene-7-methyl-1,6-octadiene, myrcene natural | |
CHEBI:17221 | |
CC(=CCCC(=C)C=C)C |
C10H16 | |
UAHWPYUMFXYFJY-UHFFFAOYSA-N | |
31253 | |
7-methyl-3-methylideneocta-1,6-diene |
Safety and Handling
GHS H Statement
Flammable liquid and vapour.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.
May produce an al
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: Immediately call a POISON CENTER o
GHS Signal Word: Danger
EINECSNumber : 204-622-5
RUO – Research Use Only