missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Myrcene, 90%, tech., stabilized, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 128080050

 View more versions of this product

Catalog No. AC128080050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Myrcene
Colorless to Yellow
165.0°C
Authentic
Glass bottle
1.4660 to 1.4720
5 mL
0.795
myrcene, beta-myrcene, 7-methyl-3-methyleneocta-1,6-diene, 7-methyl-3-methylene-1,6-octadiene, 1,6-octadiene, 7-methyl-3-methylene, beta-geraniolene, .beta.-myrcene, 2-methyl-6-methylene-2,7-octadiene, 3-methylene-7-methyl-1,6-octadiene, myrcene natural
UAHWPYUMFXYFJY-UHFFFAOYSA-N
7-methyl-3-methylideneocta-1,6-diene
Stabilized with 0.01% alpha-Tocopherol
CHEBI:17221
90%
123-35-3,127-91-3,138-86-3
0.7950g/mL
44°C
88% min. (GC)
C10H16
H2C=CHC(=CH2)CH2CH2CH=C(CH3)2
01,264
15,6416
Solubility in water: practically insoluble. Other solubilities: soluble in alcohol, chloroform, ether and, glacial acetic acid
CC(=CCCC(=C)C=C)C
136.24
31253
136.24
Liquid

Chemical Identifiers

123-35-3
136.24
myrcene, beta-myrcene, 7-methyl-3-methyleneocta-1,6-diene, 7-methyl-3-methylene-1,6-octadiene, 1,6-octadiene, 7-methyl-3-methylene, beta-geraniolene, .beta.-myrcene, 2-methyl-6-methylene-2,7-octadiene, 3-methylene-7-methyl-1,6-octadiene, myrcene natural
CHEBI:17221
CC(=CCCC(=C)C=C)C
C10H16
UAHWPYUMFXYFJY-UHFFFAOYSA-N
31253
7-methyl-3-methylideneocta-1,6-diene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapour.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.
May produce an al

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: Immediately call a POISON CENTER o

GHS Signal Word: Danger

EINECSNumber : 204-622-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only