missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific Chemicals Myristic acid, 99%

Supplier:  Thermo Scientific Chemicals 156962500

 View more versions of this product

Catalog No. AC156962500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Myristic acid
98.5
50.0°C to 57.0°C
326.0°C
Authentic
Glass bottle
CH3(CH2)12CO2H
250 g
15, 6419
Solubility in water: insoluble. Other solubilities: soluble in alcohol, methanol, ether, petr.ether,, benzene and chloroform
CCCCCCCCCCCCCC(O)=O
228.38
CHEBI:28875
99%
544-63-8
100.0
White
>110°C
98.5% min. (GC)
C14H28O2
MFCD00002744
02, 365
myristic acid, n-tetradecanoic acid, n-tetradecoic acid, crodacid, n-tetradecan-1-oic acid, 1-tridecanecarboxylic acid, hydrofol acid 1495, univol u 316s, emery 655, myristinsaeure
TUNFSRHWOTWDNC-UHFFFAOYSA-N
tetradecanoic acid
11005
228.37
Crystalline Mass, Powder, Flakes or Chunks

Chemical Identifiers

544-63-8
228.38
TUNFSRHWOTWDNC-UHFFFAOYSA-N
11005
tetradecanoic acid
C14H28O2
MFCD00002744
myristic acid, n-tetradecanoic acid, n-tetradecoic acid, crodacid, n-tetradecan-1-oic acid, 1-tridecanecarboxylic acid, hydrofol acid 1495, univol u 316s, emery 655, myristinsaeure
CHEBI:28875
CCCCCCCCCCCCCC(O)=O
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.

GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact len

GHS Signal Word: Warning

EINECSNumber : 208-875-2

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only