missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Myristoyl Chloride 98.0+%, TCI America™
$60.56 - $410.03
Chemical Identifiers
CAS | 112-64-1 |
---|---|
Molecular Formula | C14H27ClO |
Molecular Weight (g/mol) | 246.819 |
MDL Number | MFCD00000741 |
InChI Key | LPWCRLGKYWVLHQ-UHFFFAOYSA-N |
Synonym | myristoyl chloride, myristoylchloride, tetradecanoylchloride, e r eao poundae, myristic acid chloride, tetradecanoic acid chloride, acmc-2099fr, ksc181q3r |
PubChem CID | 66986 |
IUPAC Name | tetradecanoyl chloride |
SMILES | CCCCCCCCCCCCCC(=O)Cl |
Chemical Identifiers
112-64-1 | |
246.819 | |
LPWCRLGKYWVLHQ-UHFFFAOYSA-N | |
66986 | |
CCCCCCCCCCCCCC(=O)Cl |
C14H27ClO | |
MFCD00000741 | |
myristoyl chloride, myristoylchloride, tetradecanoylchloride, e r eao poundae, myristic acid chloride, tetradecanoic acid chloride, acmc-2099fr, ksc181q3r | |
tetradecanoyl chloride |
Specifications
112-64-1 | |
165°C | |
MFCD00000741 | |
3265 | |
LPWCRLGKYWVLHQ-UHFFFAOYSA-N | |
tetradecanoyl chloride | |
66986 | |
≥98.0% (GC,T) | |
Myristoyl Chloride |
Yellow | |
C14H27ClO | |
25 g | |
myristoyl chloride, myristoylchloride, tetradecanoylchloride, e r eao poundae, myristic acid chloride, tetradecanoic acid chloride, acmc-2099fr, ksc181q3r | |
CCCCCCCCCCCCCC(=O)Cl | |
246.819 | |
246.82 | |
Liquid |
Safety and Handling
EINECSNumber : (2)-0632
TSCA : Yes