missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-(1,3-benzodioxol-5-ylmethyl)-n-methylamine, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC01646DA
Specifications
| N-(1, 3-Benzodioxol-5-ylmethyl)-N-methylamine | |
| 97% | |
| C9H12NO2 | |
| 1g | |
| CEPGPPSMCRKGFJ-UHFFFAOYSA-O | |
| 1-(1,3-benzodioxol-5-yl)-N-methylmethanamine | |
| 421238 | |
| 97% |
| 15205-27-3 | |
| Amber glass bottle | |
| MFCD04496422 | |
| n-1,3-benzodioxol-5-ylmethyl-n-methylamine, 1-1,3-benzodioxol-5-yl-n-methylmethanamine, 2h-1,3-benzodioxol-5-ylmethyl methyl amine, n-methyl-3,4-methylenedioxy benzylamine, 1,3-benzodioxole-5-methanamine,n-methyl, 1,3-benzodioxole-5-methanamine, n-methyl, 2h-1,3-benzodioxol-5-yl methyl methyl amine, 1,3-benzodioxol-5-ylmethyl methylaminehydrochloride, n-methylpiperonylamine, 3,4-methylenedioxy-n-methylbenzylamine | |
| C[NH2+]CC1=CC=C2OCOC2=C1 | |
| 166.20 | |
| 165.19 |
Chemical Identifiers
| 15205-27-3 | |
| 166.20 | |
| CEPGPPSMCRKGFJ-UHFFFAOYSA-O | |
| 421238 | |
| C[NH2+]CC1=CC=C2OCOC2=C1 |
| C9H12NO2 | |
| MFCD04496422 | |
| n-1,3-benzodioxol-5-ylmethyl-n-methylamine, 1-1,3-benzodioxol-5-yl-n-methylmethanamine, 2h-1,3-benzodioxol-5-ylmethyl methyl amine, n-methyl-3,4-methylenedioxy benzylamine, 1,3-benzodioxole-5-methanamine,n-methyl, 1,3-benzodioxole-5-methanamine, n-methyl, 2h-1,3-benzodioxol-5-yl methyl methyl amine, 1,3-benzodioxol-5-ylmethyl methylaminehydrochloride, n-methylpiperonylamine, 3,4-methylenedioxy-n-methylbenzylamine | |
| 1-(1,3-benzodioxol-5-yl)-N-methylmethanamine |