missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

n-(1,3-benzodioxol-5-ylmethyl)-n-methylamine, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC01646EA

 View more versions of this product

Catalog No. CC01646EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

N-(1, 3-Benzodioxol-5-ylmethyl)-N-methylamine
97%
C9H12NO2
10g
CEPGPPSMCRKGFJ-UHFFFAOYSA-O
1-(1,3-benzodioxol-5-yl)-N-methylmethanamine
421238
97%
15205-27-3
Amber glass bottle
MFCD04496422
n-1,3-benzodioxol-5-ylmethyl-n-methylamine, 1-1,3-benzodioxol-5-yl-n-methylmethanamine, 2h-1,3-benzodioxol-5-ylmethyl methyl amine, n-methyl-3,4-methylenedioxy benzylamine, 1,3-benzodioxole-5-methanamine,n-methyl, 1,3-benzodioxole-5-methanamine, n-methyl, 2h-1,3-benzodioxol-5-yl methyl methyl amine, 1,3-benzodioxol-5-ylmethyl methylaminehydrochloride, n-methylpiperonylamine, 3,4-methylenedioxy-n-methylbenzylamine
C[NH2+]CC1=CC=C2OCOC2=C1
166.20
165.19

Chemical Identifiers

15205-27-3
166.20
CEPGPPSMCRKGFJ-UHFFFAOYSA-O
421238
C[NH2+]CC1=CC=C2OCOC2=C1
C9H12NO2
MFCD04496422
n-1,3-benzodioxol-5-ylmethyl-n-methylamine, 1-1,3-benzodioxol-5-yl-n-methylmethanamine, 2h-1,3-benzodioxol-5-ylmethyl methyl amine, n-methyl-3,4-methylenedioxy benzylamine, 1,3-benzodioxole-5-methanamine,n-methyl, 1,3-benzodioxole-5-methanamine, n-methyl, 2h-1,3-benzodioxol-5-yl methyl methyl amine, 1,3-benzodioxol-5-ylmethyl methylaminehydrochloride, n-methylpiperonylamine, 3,4-methylenedioxy-n-methylbenzylamine
1-(1,3-benzodioxol-5-yl)-N-methylmethanamine
Videos
SDS Documents

Documents

Promotions

Promotions