missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,1-Di-Fmoc-L-histidine, 95%, Thermo Scientific™

$158.47 - $158.47

Chemical Identifiers

CAS 98929-98-7
Molecular Formula C36H29N3O6
Molecular Weight (g/mol) 599.64
MDL Number MFCD00070658
InChI Key WTBXFPFCUZKREB-UHFFFAOYNA-N
Synonym fmoc-his fmoc-oh, n,n'-bis 9-fluorenylmethyloxycarbonyl-l-histidine, s-3-1-9h-fluoren-9-yl methoxy carbonyl-1h-imidazol-4-yl-2-9h-fluoren-9-yl methoxy carbonyl amino propanoic acid, l-histidine, n,1-bis 9h-fluoren-9-ylmethoxy carbonyl, fmoc-his fmoc, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-1-9h-fluoren-9-ylmethoxy carbonyl imidazol-4-yl propanoic acid, ambotzfaa1497, fmoc-l-his fmoc-oh, pubchem10020, fmoc-histidine fmoc-oh
PubChem CID 45072174
SMILES OC(=O)C(CC1=CN(C=N1)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6293206
View Documents Promotion Details
Thermo Scientific Chemicals
H6293206
5 g
Each for $158.47
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

98929-98-7
599.64
WTBXFPFCUZKREB-UHFFFAOYNA-N
45072174
C36H29N3O6
MFCD00070658
fmoc-his fmoc-oh, n,n'-bis 9-fluorenylmethyloxycarbonyl-l-histidine, s-3-1-9h-fluoren-9-yl methoxy carbonyl-1h-imidazol-4-yl-2-9h-fluoren-9-yl methoxy carbonyl amino propanoic acid, l-histidine, n,1-bis 9h-fluoren-9-ylmethoxy carbonyl, fmoc-his fmoc, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-1-9h-fluoren-9-ylmethoxy carbonyl imidazol-4-yl propanoic acid, ambotzfaa1497, fmoc-l-his fmoc-oh, pubchem10020, fmoc-histidine fmoc-oh
OC(=O)C(CC1=CN(C=N1)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

98929-98-7
MFCD00070658
fmoc-his fmoc-oh, n,n'-bis 9-fluorenylmethyloxycarbonyl-l-histidine, s-3-1-9h-fluoren-9-yl methoxy carbonyl-1h-imidazol-4-yl-2-9h-fluoren-9-yl methoxy carbonyl amino propanoic acid, l-histidine, n,1-bis 9h-fluoren-9-ylmethoxy carbonyl, fmoc-his fmoc, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-1-9h-fluoren-9-ylmethoxy carbonyl imidazol-4-yl propanoic acid, ambotzfaa1497, fmoc-l-his fmoc-oh, pubchem10020, fmoc-histidine fmoc-oh
OC(=O)C(CC1=CN(C=N1)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
599.64
599.63
N,1-Di-Fmoc-L-histidine
C36H29N3O6
5 g
WTBXFPFCUZKREB-UHFFFAOYNA-N
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[1-(9H-fluoren-9-ylmethoxycarbonyl)imidazol-4-yl]propanoic acid
45072174
95%
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Store at -20°C

SDS
missing translation for 'documents'

missing translation for 'documents'