Learn More
N-(1-Naphthyl)ethylenediamine, dihydrochloride, 98+%, ACS reagent, Thermo Scientific Chemicals
N-(1-Naphthyl)ethylenediamine, dihydrochloride, >-98.0% (on dry substance), C12H16Cl2N2, CAS Number-1465-25-4
$290.06 - $2736.20
Chemical Identifiers
CAS | 1465-25-4 |
---|---|
MDL Number | MFCD00012556 |
InChI Key | MZNYWPRCVDMOJG-UHFFFAOYSA-N |
Synonym | n-1-naphthyl ethylenediamine dihydrochloride, n1-naphthalen-1-yl ethane-1,2-diamine dihydrochloride, marshall's reagent, n-1-naphthylethylenediamine dihydrochloride, ccris 425, unii-h734599kjl, 1,2-ethanediamine, n-1-naphthalenyl-, dihydrochloride, 2-1-naphthylamino ethylamine 2hcl, bratton-marshall reagent, n-1-naphthalenyl-1,2-ethanediamine dihydrochloride |
PubChem CID | 15106 |
ChEBI | CHEBI:53452 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC423990250
|
Thermo Scientific Chemicals
423990250 |
25 g | Glass Bottle |
Each for $290.06
|
|
||||
AC423991000
|
Thermo Scientific Chemicals
423991000 |
100 g | Glass Bottle |
Each for $717.89
|
|
||||
AC423995000
|
Thermo Scientific Chemicals
423995000 |
500 g | Glass bottle |
Each for $2,736.20
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1465-25-4 | |
MZNYWPRCVDMOJG-UHFFFAOYSA-N | |
15106 |
MFCD00012556 | |
n-1-naphthyl ethylenediamine dihydrochloride, n1-naphthalen-1-yl ethane-1,2-diamine dihydrochloride, marshall's reagent, n-1-naphthylethylenediamine dihydrochloride, ccris 425, unii-h734599kjl, 1,2-ethanediamine, n-1-naphthalenyl-, dihydrochloride, 2-1-naphthylamino ethylamine 2hcl, bratton-marshall reagent, n-1-naphthalenyl-1,2-ethanediamine dihydrochloride | |
CHEBI:53452 |
Specifications
1465-25-4 | |
100.0 | |
Beige to White or Pink | |
98+% | |
C12H16Cl2N2 | |
MFCD00012556 | |
12, II, 699 | |
15, 6491 | |
Solubility in water: soluble. Other solubilities: soluble in hot water, slightly soluble in acetone and absolute alcohol | |
C1=CC=C2C(=C1)C=CC=C2NCCN.Cl.Cl | |
259.174 | |
CHEBI:53452 | |
≥98.0% (on dry substance) | |
Powder |
98.0 | |
200°C | |
Authentic | |
Glass Bottle | |
C10H7NHCH2CH2NH2·2HCl | |
25 g | |
(to sulfanilamide) Passes | |
n-1-naphthyl ethylenediamine dihydrochloride, n1-naphthalen-1-yl ethane-1,2-diamine dihydrochloride, marshall's reagent, n-1-naphthylethylenediamine dihydrochloride, ccris 425, unii-h734599kjl, 1,2-ethanediamine, n-1-naphthalenyl-, dihydrochloride, 2-1-naphthylamino ethylamine 2hcl, bratton-marshall reagent, n-1-naphthalenyl-1,2-ethanediamine dihydrochloride | |
MZNYWPRCVDMOJG-UHFFFAOYSA-N | |
N'-naphthalen-1-ylethane-1,2-diamine;dihydrochloride | |
15106 | |
259.18 | |
ACS Reagent | |
N-(1-Naphthyl)ethylenediamine, dihydrochloride |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue
GHS Signal Word: Warning
EINECSNumber : 215-981-2
RUO – Research Use Only