missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-(1-Naphthyl)ethylenediamine Dihydrochloride, ACS, Spectrum™ Chemical
$252.50 - $37414.76
Chemical Identifiers
CAS | 1465-25-4 |
---|---|
Molecular Formula | C12H16Cl2N2 |
Molecular Weight (g/mol) | 259.17 |
InChI Key | MZNYWPRCVDMOJG-UHFFFAOYSA-N |
IUPAC Name | dihydrogen N1-(naphthalen-1-yl)ethane-1,2-diamine dichloride |
SMILES | [H+].[H+].[Cl-].[Cl-].NCCNC1=C2C=CC=CC2=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18611844
|
Spectrum Chemical Mfg Cor
N10185GM |
5 g | Amber Glass Bottle |
Each for $252.50
|
|
||||
18611845
|
Spectrum Chemical Mfg Cor
N101825GM |
25 g | Amber Glass Bottle |
Each for $676.47
|
|
||||
18611846
|
Spectrum Chemical Mfg Cor
N1018100GM |
100 g | Amber Glass Bottle |
Each for $1,911.47
|
|
||||
18611847
|
Spectrum Chemical Mfg Cor
N101810KGBL |
10 kg | Poly Pail |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
1465-25-4 | |
259.17 | |
dihydrogen N1-(naphthalen-1-yl)ethane-1,2-diamine dichloride |
C12H16Cl2N2 | |
MZNYWPRCVDMOJG-UHFFFAOYSA-N | |
[H+].[H+].[Cl-].[Cl-].NCCNC1=C2C=CC=CC2=CC=C1 |
Specifications
1465-25-4 | |
Amber Glass Bottle | |
5 g | |
[H+].[H+].[Cl-].[Cl-].NCCNC1=C2C=CC=CC2=CC=C1 | |
259.17 |
1 | |
C12H16Cl2N2 | |
MZNYWPRCVDMOJG-UHFFFAOYSA-N | |
dihydrogen N1-(naphthalen-1-yl)ethane-1,2-diamine dichloride | |
ACS |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.