missing translation for 'onlineSavingsMsg'
Learn More

N,2,3-Trimethyl-2-isopropylbutamide, Thermo Scientific™

Chemical Identifiers

CAS 51115-67-4
Molecular Formula C10H21NO
Molecular Weight (g/mol) 171.28
MDL Number MFCD00130070
InChI Key RWAXQWRDVUOOGG-UHFFFAOYSA-N
Synonym 2-isopropyl-n,2,3-trimethylbutyramide, 2-isopropyl-n,2,3-trimethylbutanamide, trimethyl isopropyl butanamide, methyl diisopropyl propionamide, n,2,3-trimethyl-2-1-methylethyl butanamide, unii-6qop5a9489, n,2,3-trimethyl-2-isopropylbutanamide, n,2,3-trimethyl-2-isopropylbutamide, butanamide, n,2,3-trimethyl-2-1-methylethyl, n,2,3-trimethyl-2-propan-2-yl butanamide
PubChem CID 65300
IUPAC Name N,2,3-trimethyl-2-propan-2-ylbutanamide
SMILES CNC(=O)C(C)(C(C)C)C(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC421930050
SDS View Documents
Thermo Scientific Chemicals
421930050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC421930250
SDS View Documents
Thermo Scientific Chemicals
421930250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

51115-67-4
171.28
RWAXQWRDVUOOGG-UHFFFAOYSA-N
65300
CNC(=O)C(C)(C(C)C)C(C)C
C10H21NO
MFCD00130070
2-isopropyl-n,2,3-trimethylbutyramide, 2-isopropyl-n,2,3-trimethylbutanamide, trimethyl isopropyl butanamide, methyl diisopropyl propionamide, n,2,3-trimethyl-2-1-methylethyl butanamide, unii-6qop5a9489, n,2,3-trimethyl-2-isopropylbutanamide, n,2,3-trimethyl-2-isopropylbutamide, butanamide, n,2,3-trimethyl-2-1-methylethyl, n,2,3-trimethyl-2-propan-2-yl butanamide
N,2,3-trimethyl-2-propan-2-ylbutanamide

Specifications

51115-67-4
C10H21NO
5g
2-isopropyl-n,2,3-trimethylbutyramide, 2-isopropyl-n,2,3-trimethylbutanamide, trimethyl isopropyl butanamide, methyl diisopropyl propionamide, n,2,3-trimethyl-2-1-methylethyl butanamide, unii-6qop5a9489, n,2,3-trimethyl-2-isopropylbutanamide, n,2,3-trimethyl-2-isopropylbutamide, butanamide, n,2,3-trimethyl-2-1-methylethyl, n,2,3-trimethyl-2-propan-2-yl butanamide
CNC(=O)C(C)(C(C)C)C(C)C
171.28
171.28
Glass bottle
MFCD00130070
15,9887
RWAXQWRDVUOOGG-UHFFFAOYSA-N
N,2,3-trimethyl-2-propan-2-ylbutanamide
65300
N, 2, 3-Trimethyl-2-isopropylbutamide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face,hands and any exposed skin thoroughly after handling.

GHS Signal Word: Warning

EINECSNumber : 256-974-4

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only