missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-(2-Aminoethyl)acetamide, 90%, Thermo Scientific™

Chemical Identifiers

CAS 1001-53-2
Molecular Formula C4H10N2O
Molecular Weight (g/mol) 102.14
MDL Number MFCD00008163
InChI Key DAKZISABEDGGSV-UHFFFAOYSA-N
Synonym n-2-aminoethyl acetamide, n-acetylethylenediamine, acetamide, n-2-aminoethyl, n-2-amino-ethyl-acetamide, unii-fql33v3kzd, fql33v3kzd, acetylethylenediamine, 2-acetamidoethylamine, acetylethylene diamine, n-acetylethylendiamine
PubChem CID 66082
IUPAC Name N-(2-aminoethyl)acetamide
SMILES CC(=O)NCCN
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC223170050
SDS View Documents
Thermo Scientific Chemicals
223170050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1001-53-2
102.14
DAKZISABEDGGSV-UHFFFAOYSA-N
66082
CC(=O)NCCN
C4H10N2O
MFCD00008163
n-2-aminoethyl acetamide, n-acetylethylenediamine, acetamide, n-2-aminoethyl, n-2-amino-ethyl-acetamide, unii-fql33v3kzd, fql33v3kzd, acetylethylenediamine, 2-acetamidoethylamine, acetylethylene diamine, n-acetylethylendiamine
N-(2-aminoethyl)acetamide

Specifications

1001-53-2
100.0
>110°C
Glass bottle
CH3C(O)NH(CH2)2NH2
5g
n-2-aminoethyl acetamide, n-acetylethylenediamine, acetamide, n-2-aminoethyl, n-2-amino-ethyl-acetamide, unii-fql33v3kzd, fql33v3kzd, acetylethylenediamine, 2-acetamidoethylamine, acetylethylene diamine, n-acetylethylendiamine
CC(=O)NCCN
102.14
102.14
N-(2-Aminoethyl)acetamide
88.0
143.0°C (10.0 mmHg)
88% min. (GC)
C4H10N2O
MFCD00008163
04, III, 522
DAKZISABEDGGSV-UHFFFAOYSA-N
N-(2-aminoethyl)acetamide
66082
90%
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only