missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-(2-Bromoethyl)phthalimide, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 106971000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
N-(2-Bromoethyl)phthalimide | |
574-98-1 | |
100.0 | |
Beige or Pink to White | |
96% min. (on dry substance) (HPLC) | |
C10H8BrNO2 | |
100 g | |
n-2-bromoethyl phthalimide, 2-2-bromoethyl isoindoline-1,3-dione, 1h-isoindole-1,3 2h-dione, 2-2-bromoethyl, 2-2-bromoethyl-1h-isoindole-1,3 2h-dione, beta-bromoethylphthalimide, 2-bromoethyl phthalimide, 1-bromo-2-phthalimidoethane, beta-phthalimidoethyl bromide, n-2-bromoethyl-phthalimide, 2-2-bromo-ethyl-isoindole-1,3-dione | |
CHZXTOCAICMPQR-UHFFFAOYSA-N | |
2-(2-bromoethyl)isoindole-1,3-dione | |
11325 | |
97% |
99+% | |
96.0 | |
81.0°C to 84.0°C | |
Authentic | |
Glass bottle | |
MFCD00005902 | |
21,461 | |
Solubility in water: insoluble in water. Other solubilities: soluble in organic solvents ca.10g/l,hardly soluble in ethanol and ether | |
C1=CC=C2C(=C1)C(=O)N(C2=O)CCBr | |
254.08 | |
254.08 | |
Crystalline Powder |
Chemical Identifiers
574-98-1 | |
254.08 | |
CHZXTOCAICMPQR-UHFFFAOYSA-N | |
11325 | |
C1=CC=C2C(=C1)C(=O)N(C2=O)CCBr |
C10H8BrNO2 | |
MFCD00005902 | |
n-2-bromoethyl phthalimide, 2-2-bromoethyl isoindoline-1,3-dione, 1h-isoindole-1,3 2h-dione, 2-2-bromoethyl, 2-2-bromoethyl-1h-isoindole-1,3 2h-dione, beta-bromoethylphthalimide, 2-bromoethyl phthalimide, 1-bromo-2-phthalimidoethane, beta-phthalimidoethyl bromide, n-2-bromoethyl-phthalimide, 2-2-bromo-ethyl-isoindole-1,3-dione | |
2-(2-bromoethyl)isoindole-1,3-dione |
Safety and Handling
EINECSNumber : 209-379-9
TSCA : TSCA
RUO – Research Use Only