missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Chloroethyl)acetamide, 98%

CAS: 7355-58-0 | C4H8ClNO | 121.57 g/mol

$98.56 - $98.56

Chemical Identifiers

CAS 7355-58-0
Molecular Formula C4H8ClNO
Molecular Weight (g/mol) 121.57
MDL Number MFCD00000965
InChI Key HSKNJSHFPPHTAQ-UHFFFAOYSA-N
Synonym n-2-chloroethyl acetamide, acetamide, n-2-chloroethyl, n-2-chloro-ethyl-acetamide, n-acetyl-2-chloroethylamine, n 2-chloroethyl acetamide, n-acetyl-ss-chlorathylamin, acmc-1bi05, n-2-chloroethyl-acetamide, 4-04-00-00449 beilstein handbook reference
PubChem CID 81815
IUPAC Name N-(2-chloroethyl)acetamide
SMILES CC(=O)NCCCl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC184520100
SDS View Documents
Thermo Scientific Chemicals
184520100
10 g
Each for $98.56
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7355-58-0
121.57
HSKNJSHFPPHTAQ-UHFFFAOYSA-N
81815
CC(=O)NCCCl
C4H8ClNO
MFCD00000965
n-2-chloroethyl acetamide, acetamide, n-2-chloroethyl, n-2-chloro-ethyl-acetamide, n-acetyl-2-chloroethylamine, n 2-chloroethyl acetamide, n-acetyl-ss-chlorathylamin, acmc-1bi05, n-2-chloroethyl-acetamide, 4-04-00-00449 beilstein handbook reference
N-(2-chloroethyl)acetamide

Specifications

7355-58-0
100.0
1.2000g/mL
Authentic
Glass bottle
1.4770 to 1.48
MFCD00000965
1.2
HSKNJSHFPPHTAQ-UHFFFAOYSA-N
N-(2-chloroethyl)acetamide
81815
98%
N-(2-Chloroethyl)acetamide, 98%
97.5
Colorless to Yellow
110°C
97.5% min. (GC)
C4H8ClNO
CH3CONHCH2CH2Cl
10 g
n-2-chloroethyl acetamide, acetamide, n-2-chloroethyl, n-2-chloro-ethyl-acetamide, n-acetyl-2-chloroethylamine, n 2-chloroethyl acetamide, n-acetyl-ss-chlorathylamin, acmc-1bi05, n-2-chloroethyl-acetamide, 4-04-00-00449 beilstein handbook reference
CC(=O)NCCCl
121.57
121.57
Viscous Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 230-884-5

RTECSNumber : AB5460000

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only