missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Hydroxyethyl)ethylenediamine-N,N',N'-triacetic Acid 98.0+%, TCI America™

$57.79 - $436.19

Chemical Identifiers

CAS 150-39-0
Molecular Formula C10H15N2Na3O7
Molecular Weight (g/mol) 344.21
MDL Number MFCD00004294
InChI Key WHNXAQZPEBNFBC-UHFFFAOYSA-K
Synonym hedta, heedta, versenol, detarol, n-2-hydroxyethyl ethylenediaminetriacetic acid, chel dm acid, versenol 120, hamp-ol acid, 2-hydroxyethyl ethylenediaminetriacetic acid, oxyethylethylenediaminetriacetic acid
PubChem CID 8773
IUPAC Name trisodium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(2-hydroxyethyl)amino)acetate
SMILES [Na+].[Na+].[Na+].OCCN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H024325G
View Documents
TCI America
H024325G
25 g N/A
Only null left
 
H0243100G
View Documents
TCI America
H0243100G
100 g N/A
Only null left
 
H0243500G
View Documents
TCI America
H0243500G
500 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

150-39-0
344.21
WHNXAQZPEBNFBC-UHFFFAOYSA-K
8773
[Na+].[Na+].[Na+].OCCN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O
C10H15N2Na3O7
MFCD00004294
hedta, heedta, versenol, detarol, n-2-hydroxyethyl ethylenediaminetriacetic acid, chel dm acid, versenol 120, hamp-ol acid, 2-hydroxyethyl ethylenediaminetriacetic acid, oxyethylethylenediaminetriacetic acid
trisodium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(2-hydroxyethyl)amino)acetate

Specifications

150-39-0
C10H15N2Na3O7
25 g
WHNXAQZPEBNFBC-UHFFFAOYSA-K
trisodium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(2-hydroxyethyl)amino)acetate
8773
≥98.0% (T)
N-(2-Hydroxyethyl)ethylenediamine-N,N′,N′-triacetic Acid
White
MFCD00004294
hedta, heedta, versenol, detarol, n-2-hydroxyethyl ethylenediaminetriacetic acid, chel dm acid, versenol 120, hamp-ol acid, 2-hydroxyethyl ethylenediaminetriacetic acid, oxyethylethylenediaminetriacetic acid
[Na+].[Na+].[Na+].OCCN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O
344.21
278.26
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

RTECSNumber : MB9185000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'