missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-(2-Hydroxyethyl)piperazine, 98.5%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 121191000

 View more versions of this product

Catalog No. AC121191000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

N-(2-Hydroxyethyl)piperazine
103-76-4
100.0
Colorless to Yellow
245°C to 246°C
Authentic
Glass bottle
1.5055 to 1.5075
100 g
1.06
Solubility in water: miscible with water. Other solubilities: miscible with most common organic solvents
C1CN(CCN1)CCO
130.19
130.19
Oily Liquid
98.5%
98.0
-38.5°C
1.0600g/mL
135°C
98% min. (GC)
C6H14N2O
MFCD00005970
23, II, 6
n-2-hydroxyethyl piperazine, 1-piperazineethanol, 1-2-hydroxyethyl piperazine, hydroxyethylpiperazine, 2-1-piperazinyl ethanol, 2-hydroxyethylpiperazine, 2-piperazinoethanol, 2-piperazin-1-yl ethan-1-ol, 1-piperazinethanol, 4-2-hydroxyethyl piperazine
WFCSWCVEJLETKA-UHFFFAOYSA-N
2-piperazin-1-ylethanol
7677
98.50%

Chemical Identifiers

103-76-4
130.19
WFCSWCVEJLETKA-UHFFFAOYSA-N
7677
C1CN(CCN1)CCO
C6H14N2O
MFCD00005970
n-2-hydroxyethyl piperazine, 1-piperazineethanol, 1-2-hydroxyethyl piperazine, hydroxyethylpiperazine, 2-1-piperazinyl ethanol, 2-hydroxyethylpiperazine, 2-piperazinoethanol, 2-piperazin-1-yl ethan-1-ol, 1-piperazinethanol, 4-2-hydroxyethyl piperazine
2-piperazin-1-ylethanol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 203-142-3

RTECSNumber : TL6825000

TSCA : TSCA

Recommended Storage : Yellowing of the substance during storage possible

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only