missing translation for 'onlineSavingsMsg'
Learn More

N-(3-Acetyl-4-hydroxyphenyl)butyramide, 97%

CAS: 40188-45-2 | C12H15NO3 | 221.26 g/mol

$67.60 - $773.21

Chemical Identifiers

CAS 40188-45-2
Molecular Formula C12H15NO3
Molecular Weight (g/mol) 221.26
MDL Number MFCD00798556
InChI Key FGWZEOPEZISTTR-UHFFFAOYSA-N
Synonym 2-acetyl-4-butyramidophenol, n-3-acetyl-4-hydroxyphenyl butanamide, n-3-acetyl-4-hydroxyphenyl butyramide, ac-hophe-oh, unii-14qjb65r4v, 3'-acetyl-4'-hydroxybutyranilide, butanamide, n-3-acetyl-4-hydroxyphenyl, 5'-butyramido-2'-hydroxyacetophenone, acebutolol impurity c, n-3-acetyl-4-hydroxyphenyl butanamid
PubChem CID 736331
IUPAC Name N-(3-acetyl-4-hydroxyphenyl)butanamide
SMILES CCCC(=O)NC1=CC=C(O)C(=C1)C(C)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1867218
View Documents
Thermo Scientific Chemicals
A1867218
50 g
Each for $67.60
Only null left
 
AAA186720B
View Documents
Thermo Scientific Chemicals
A186720B
1000 g
Each for $773.21
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

40188-45-2
221.26
FGWZEOPEZISTTR-UHFFFAOYSA-N
736331
CCCC(=O)NC1=CC=C(O)C(=C1)C(C)=O
C12H15NO3
MFCD00798556
2-acetyl-4-butyramidophenol, n-3-acetyl-4-hydroxyphenyl butanamide, n-3-acetyl-4-hydroxyphenyl butyramide, ac-hophe-oh, unii-14qjb65r4v, 3'-acetyl-4'-hydroxybutyranilide, butanamide, n-3-acetyl-4-hydroxyphenyl, 5'-butyramido-2'-hydroxyacetophenone, acebutolol impurity c, n-3-acetyl-4-hydroxyphenyl butanamid
N-(3-acetyl-4-hydroxyphenyl)butanamide

Specifications

40188-45-2
C12H15NO3
50 g
FGWZEOPEZISTTR-UHFFFAOYSA-N
N-(3-acetyl-4-hydroxyphenyl)butanamide
736331
97%
124°C to 126°C
MFCD00798556
2-acetyl-4-butyramidophenol, n-3-acetyl-4-hydroxyphenyl butanamide, n-3-acetyl-4-hydroxyphenyl butyramide, ac-hophe-oh, unii-14qjb65r4v, 3'-acetyl-4'-hydroxybutyranilide, butanamide, n-3-acetyl-4-hydroxyphenyl, 5'-butyramido-2'-hydroxyacetophenone, acebutolol impurity c, n-3-acetyl-4-hydroxyphenyl butanamid
CCCC(=O)NC1=CC=C(O)C(=C1)C(C)=O
221.26
221.26
N-(3-Acetyl-4-hydroxyphenyl)butyramide
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 254-831-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only