missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-(4-Aminobutyl)-2-naphthalenesulfonamide Hydrochloride 98.0+%, TCI America™
$38.06 - $351.21
Chemical Identifiers
CAS | 89108-46-3 |
---|---|
Molecular Formula | C14H19ClN2O2S |
Molecular Weight (g/mol) | 314.828 |
MDL Number | MFCD00058052 |
InChI Key | WYFVKUWCEVMLOL-UHFFFAOYSA-N |
Synonym | W-12 Hydrochloride |
PubChem CID | 9839864 |
IUPAC Name | N-(4-aminobutyl)naphthalene-2-sulfonamide;hydrochloride |
SMILES | C1=CC=C2C=C(C=CC2=C1)S(=O)(=O)NCCCCN.Cl |
Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
A240825MG
|
TCI America
A240825MG |
25 mg |
Each for $38.06
|
|
|||||
A2408500MG
|
TCI America
A2408500MG |
500 mg |
Each for $351.21
|
|
|||||
Chemical Identifiers
89108-46-3 | |
314.828 | |
WYFVKUWCEVMLOL-UHFFFAOYSA-N | |
9839864 | |
C1=CC=C2C=C(C=CC2=C1)S(=O)(=O)NCCCCN.Cl |
C14H19ClN2O2S | |
MFCD00058052 | |
W-12 Hydrochloride | |
N-(4-aminobutyl)naphthalene-2-sulfonamide;hydrochloride |
Specifications
89108-46-3 | |
White-Yellow | |
MFCD00058052 | |
W-12 Hydrochloride | |
C1=CC=C2C=C(C=CC2=C1)S(=O)(=O)NCCCCN.Cl | |
314.828 | |
314.83 | |
Crystalline Powder |
183°C | |
C14H19ClN2O2S | |
25 mg | |
WYFVKUWCEVMLOL-UHFFFAOYSA-N | |
N-(4-aminobutyl)naphthalene-2-sulfonamide;hydrochloride | |
9839864 | |
≥98.0% (HPLC,N) | |
N-(4-Aminobutyl)-2-naphthalenesulfonamide Hydrochloride |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator