Learn More
N-(4-Ethylphenyl)maleimide, 98+%, Thermo Scientific Chemicals
$101.64 - $364.77
Chemical Identifiers
CAS | 76620-00-3 |
---|---|
Molecular Formula | C12H11NO2 |
Molecular Weight (g/mol) | 201.225 |
MDL Number | MFCD00014538 |
InChI Key | FHVHFKZQDVQILM-UHFFFAOYSA-N |
Synonym | n-4-ethylphenyl maleimide, 1-4-ethylphenyl-1h-pyrrole-2,5-dione, 1-4-ethylphenyl pyrrole-2,5-dione, 1-4-ethylphenyl maleimide, 1-4-ethylphenyl-2,5-dihydro-1h-pyrrole-2,5-dione, 1-4-ethylphenyl azoline-2,5-dione, 1-4-ethylphenyl-1h-pyrrole-2,5-dione #, 1h-pyrrole-2,5-dione, 1-4-ethylphenyl |
PubChem CID | 522624 |
IUPAC Name | 1-(4-ethylphenyl)pyrrole-2,5-dione |
SMILES | CCC1=CC=C(C=C1)N2C(=O)C=CC2=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0105804
|
Thermo Scientific Chemicals
L0105804 |
2 g |
Each for $101.64
|
|
|||||
AAL0105809
|
Thermo Scientific Chemicals
L0105809 |
10 g |
Each for $364.77
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
76620-00-3 | |
201.225 | |
FHVHFKZQDVQILM-UHFFFAOYSA-N | |
522624 | |
CCC1=CC=C(C=C1)N2C(=O)C=CC2=O |
C12H11NO2 | |
MFCD00014538 | |
n-4-ethylphenyl maleimide, 1-4-ethylphenyl-1h-pyrrole-2,5-dione, 1-4-ethylphenyl pyrrole-2,5-dione, 1-4-ethylphenyl maleimide, 1-4-ethylphenyl-2,5-dihydro-1h-pyrrole-2,5-dione, 1-4-ethylphenyl azoline-2,5-dione, 1-4-ethylphenyl-1h-pyrrole-2,5-dione #, 1h-pyrrole-2,5-dione, 1-4-ethylphenyl | |
1-(4-ethylphenyl)pyrrole-2,5-dione |
Specifications
76620-00-3 | |
C12H11NO2 | |
2 g | |
n-4-ethylphenyl maleimide, 1-4-ethylphenyl-1h-pyrrole-2,5-dione, 1-4-ethylphenyl pyrrole-2,5-dione, 1-4-ethylphenyl maleimide, 1-4-ethylphenyl-2,5-dihydro-1h-pyrrole-2,5-dione, 1-4-ethylphenyl azoline-2,5-dione, 1-4-ethylphenyl-1h-pyrrole-2,5-dione #, 1h-pyrrole-2,5-dione, 1-4-ethylphenyl | |
CCC1=CC=C(C=C1)N2C(=O)C=CC2=O | |
201.225 | |
201.22 | |
N-(4-Ethylphenyl)maleimide |
64°C to 67°C | |
MFCD00014538 | |
7022079 | |
FHVHFKZQDVQILM-UHFFFAOYSA-N | |
1-(4-ethylphenyl)pyrrole-2,5-dione | |
522624 | |
≥98% |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only