missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-(4-Fluorobenzyl)-2-methylaniline, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals H56891MD

 View more versions of this product

Catalog No. AAH56891MD


Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

N-(4-Fluorobenzyl)-2-methylaniline
C14H14FN
250 mg
JEDDLDDCZKZPOX-UHFFFAOYSA-N
N-[(4-fluorophenyl)methyl]-2-methylaniline
28489996
97%
1020963-78-3
MFCD00454861
n-4-fluorobenzyl-2-methylaniline, n-4-fluorophenyl methyl-2-methylaniline
CC1=CC=CC=C1NCC2=CC=C(C=C2)F
215.271
215.27

Chemical Identifiers

1020963-78-3
215.271
JEDDLDDCZKZPOX-UHFFFAOYSA-N
28489996
CC1=CC=CC=C1NCC2=CC=C(C=C2)F
C14H14FN
MFCD00454861
n-4-fluorobenzyl-2-methylaniline, n-4-fluorophenyl methyl-2-methylaniline
N-[(4-fluorophenyl)methyl]-2-methylaniline
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions