Learn More
N-(5-chloro-2-pyridyl)bis(trifluoromethanesulfonimide), Thermo Scientific™
Supplier: Thermo Scientific Chemicals 378800250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Coupling reagentSpecifications
N-(5-Chloro-2-pyridyl)bis(trifluoromethanesulfonimide) | |
77.0°C to 90.0°C (0.2mmHg) | |
C7H3ClF6N2O4S2 | |
25g | |
TUFGVZMNGTYAQD-UHFFFAOYSA-N | |
N-(5-chloropyridin-2-yl)-1,1,1-trifluoro-N-(trifluoromethylsulfonyl)methanesulfonamide | |
388544 |
145100-51-2 | |
Glass Bottle | |
MFCD00191833 | |
2-n,n-bis trifluoromethylsulphonyl amino-5-chloropyridine, 2-n,n-bis trifluoromethanesulfonyl amino-5-chloropyridine, comins' reagent, n-5-chloro-2-pyridyl triflimide, 2-n,n-bis trifluoromethylsulfonyl amino-5-chloropyridine, comins triflating reagent, n-5-chloro-2-pyridyl bis trifluoromethanesulfonimide, n-5-chloropyridin-2-yl-1,1,1-trifluoro-n-trifluoromethylsulfonyl methanesulfonamide, n-5-chloropyridin-2-yl triflimide, n-5-chloropyridin-2-yl-1,1,1-trifluoro-n-trifluoromethyl sulfonyl methanesulfonamide | |
C1=CC(=NC=C1Cl)N(S(=O)(=O)C(F)(F)F)S(=O)(=O)C(F)(F)F | |
392.69 | |
392.69 |
Chemical Identifiers
145100-51-2 | |
392.69 | |
TUFGVZMNGTYAQD-UHFFFAOYSA-N | |
388544 | |
C1=CC(=NC=C1Cl)N(S(=O)(=O)C(F)(F)F)S(=O)(=O)C(F)(F)F |
C7H3ClF6N2O4S2 | |
MFCD00191833 | |
2-n,n-bis trifluoromethylsulphonyl amino-5-chloropyridine, 2-n,n-bis trifluoromethanesulfonyl amino-5-chloropyridine, comins' reagent, n-5-chloro-2-pyridyl triflimide, 2-n,n-bis trifluoromethylsulfonyl amino-5-chloropyridine, comins triflating reagent, n-5-chloro-2-pyridyl bis trifluoromethanesulfonimide, n-5-chloropyridin-2-yl-1,1,1-trifluoro-n-trifluoromethylsulfonyl methanesulfonamide, n-5-chloropyridin-2-yl triflimide, n-5-chloropyridin-2-yl-1,1,1-trifluoro-n-trifluoromethyl sulfonyl methanesulfonamide | |
N-(5-chloropyridin-2-yl)-1,1,1-trifluoro-N-(trifluoromethylsulfonyl)methanesulfonamide |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protec
GHS Signal Word: Warning
RUO â Research Use Only