missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-(5-Hexynyl)phthalimide, 97%, Thermo Scientific Chemicals
$142.18 - $566.56
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH5348906
|
Thermo Scientific Chemicals
H5348906 |
5 g |
Each for $142.18
|
|
|||||
AAH5348914
|
Thermo Scientific Chemicals
H5348914 |
25 g |
Each for $566.56
|
|
|||||
Description
6-amino-1-hexyne was prepared from N-(5-Hexynyl)phthalimide which is the starting material. Used in the synthesis of Amino-alkyne.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications6-amino-1-hexyne was prepared from N-(5-Hexynyl)phthalimide which is the starting material. Used in the synthesis of Amino-alkyne.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
6097-08-1 | |
MFCD00671372 |
Specifications
6097-08-1 | |
C10H10O3 | |
5 g | |
LZPNXAULYJPXEH-AATRIKPKSA-N | |
(E)-3-(3-methoxyphenyl)prop-2-enoic acid | |
637668 | |
97% |
68°C to 72°C | |
MFCD00671372 | |
3-methoxycinnamic acid, 3-3-methoxyphenyl acrylic acid, 2e-3-3-methoxyphenyl acrylic acid, 2e-3-3-methoxyphenyl prop-2-enoic acid, trans-3-methoxycinnamic acid, m-methoxycinnamic acid, trans-3-3-methoxyphenyl acrylic acid, e-3-3-methoxyphenyl acrylic acid, 2-propenoic acid, 3-3-methoxyphenyl-, 2e, trans-m-methoxycinnamic acid | |
COC1=CC=CC(=C1)C=CC(=O)O | |
178.19 | |
227.26 | |
N-(5-Hexynyl)phthalimide |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only