missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-(6-Aminohexyl)-1-naphthalenesulfonamide Hydrochloride 98.0+%, TCI America™
$43.25 - $418.68
Chemical Identifiers
CAS | 61714-25-8 |
---|---|
Molecular Formula | C16H23ClN2O2S |
Molecular Weight (g/mol) | 342.88 |
MDL Number | MFCD00058043 |
InChI Key | HOCSVIGHWPLMFC-UHFFFAOYSA-N |
Synonym | w-5 hydrochloride, n-6-aminohexyl-1-naphthalenesulfonamide hydrochloride, w-5, hydrochloride, n-6-aminohexyl naphthalene-1-sulfonamide hydrochloride, n-6-aminohexyl-1-naphthalenesulphonamide hydrochloride, n-6-aminohexyl-1-naphthalenesulfonamide hcl, n-6-aminohexyl-1-naphthalenesulfonamide, hcl, acmc-209mvm, n-6-aminohexyl-1-naphthalenesulfonamide monohydrochlorde, 1-naphthalenesulfonamide, n-6-aminohexyl-, monohydrochlorde |
PubChem CID | 173829 |
IUPAC Name | hydrogen N-(6-aminohexyl)naphthalene-1-sulfonamide chloride |
SMILES | [H+].[Cl-].NCCCCCCNS(=O)(=O)C1=C2C=CC=CC2=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
A241025MG
|
TCI America
A241025MG |
25 mg |
Each for $43.25
|
|
|||||
A2410500MG
|
TCI America
A2410500MG |
500 mg |
Each for $418.68
|
|
|||||
Chemical Identifiers
61714-25-8 | |
342.88 | |
HOCSVIGHWPLMFC-UHFFFAOYSA-N | |
173829 | |
[H+].[Cl-].NCCCCCCNS(=O)(=O)C1=C2C=CC=CC2=CC=C1 |
C16H23ClN2O2S | |
MFCD00058043 | |
w-5 hydrochloride, n-6-aminohexyl-1-naphthalenesulfonamide hydrochloride, w-5, hydrochloride, n-6-aminohexyl naphthalene-1-sulfonamide hydrochloride, n-6-aminohexyl-1-naphthalenesulphonamide hydrochloride, n-6-aminohexyl-1-naphthalenesulfonamide hcl, n-6-aminohexyl-1-naphthalenesulfonamide, hcl, acmc-209mvm, n-6-aminohexyl-1-naphthalenesulfonamide monohydrochlorde, 1-naphthalenesulfonamide, n-6-aminohexyl-, monohydrochlorde | |
hydrogen N-(6-aminohexyl)naphthalene-1-sulfonamide chloride |
Specifications
61714-25-8 | |
White-Yellow | |
MFCD00058043 | |
w-5 hydrochloride, n-6-aminohexyl-1-naphthalenesulfonamide hydrochloride, w-5, hydrochloride, n-6-aminohexyl naphthalene-1-sulfonamide hydrochloride, n-6-aminohexyl-1-naphthalenesulphonamide hydrochloride, n-6-aminohexyl-1-naphthalenesulfonamide hcl, n-6-aminohexyl-1-naphthalenesulfonamide, hcl, acmc-209mvm, n-6-aminohexyl-1-naphthalenesulfonamide monohydrochlorde, 1-naphthalenesulfonamide, n-6-aminohexyl-, monohydrochlorde | |
[H+].[Cl-].NCCCCCCNS(=O)(=O)C1=C2C=CC=CC2=CC=C1 | |
342.88 | |
342.88 | |
Crystalline Powder |
164°C | |
C16H23ClN2O2S | |
25 mg | |
HOCSVIGHWPLMFC-UHFFFAOYSA-N | |
hydrogen N-(6-aminohexyl)naphthalene-1-sulfonamide chloride | |
173829 | |
≥98.0% (HPLC,N) | |
N-(6-Aminohexyl)-1-naphthalenesulfonamide Hydrochloride |
Safety and Handling
TSCA : No
Recommended Storage : Freezer