missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-(6-Bromohexyl)phthalimide, 97%, Thermo Scientific Chemicals

$189.92 - $2969.37

Chemical Identifiers

CAS 24566-79-8
Molecular Formula C14H16BrNO2
Molecular Weight (g/mol) 310.191
MDL Number MFCD00023098
InChI Key OAZFTIPKNPTDIO-UHFFFAOYSA-N
PubChem CID 141120
IUPAC Name 2-(6-bromohexyl)isoindole-1,3-dione
SMILES C1=CC=C2C(=C1)C(=O)N(C2=O)CCCCCCBr
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2512806
View Documents
Thermo Scientific Chemicals
B2512806
5 g
Each for $189.92
 
AAB2512814
View Documents
Thermo Scientific Chemicals
B2512814
25 g
Each for $827.26
 
AAB2512822
View Documents
Thermo Scientific Chemicals
B2512822
100 g
Each for $2,969.37
 
Description

Description

N-(6-Bromohexyl)phthalimide is used as a pharmaceutical intermediate and in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-(6-Bromohexyl)phthalimide is used as a pharmaceutical intermediate and in organic synthesis.

Solubility
Insoluble in water.

Notes
Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

24566-79-8
310.191
OAZFTIPKNPTDIO-UHFFFAOYSA-N
2-(6-bromohexyl)isoindole-1,3-dione
C14H16BrNO2
MFCD00023098
141120
C1=CC=C2C(=C1)C(=O)N(C2=O)CCCCCCBr

Specifications

24566-79-8
186°C to 188°C (0.4 mmHg)
C14H16BrNO2
5 g
n-6-bromohexyl phthalimide, 2-6-bromohexyl isoindoline-1,3-dione, n-6-bromohexyl-phthalimide, 2-6-bromohexyl-1h-isoindole-1,3 2h-dione, 2-6-bromohexyl isoindole-1,3-dione, 2-6-bromohexyl-2,3-dihydro-1h-isoindole-1,3-dione, 2-6-bromo-hexyl-isoindole-1,3-dione, 1h-isoindole-1,3 2h-dione,2-6-bromohexyl, 1h-isoindole-1,3 2h-dione, 2-6-bromohexyl, 6-n-phthalimido hexyl bromide
OAZFTIPKNPTDIO-UHFFFAOYSA-N
2-(6-bromohexyl)isoindole-1,3-dione
141120
97%
54°C to 60°C
Odorless
MFCD00023098
186875
Insoluble in water.
C1=CC=C2C(=C1)C(=O)N(C2=O)CCCCCCBr
310.191
310.19
N-(6-Bromohexyl)phthalimide
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only