Learn More
N-(9-Fluorenylmethoxycarbonyloxy)succinimide, 98%, Thermo Scientific Chemicals
$54.90 - $542.93
Chemical Identifiers
CAS | 82911-69-1 |
---|---|
Molecular Formula | C19H15NO5 |
Molecular Weight (g/mol) | 337.331 |
MDL Number | MFCD00010733 |
InChI Key | WMSUFWLPZLCIHP-UHFFFAOYSA-N |
Synonym | fmoc-osu, n-9-fluorenylmethoxycarbonyloxy succinimide, fmoc-onsu, 9-fluorenylmethyl succinimidyl carbonate, fmoc n-hydroxysuccinimide ester, 9h-fluoren-9-yl methyl 2,5-dioxopyrrolidin-1-yl carbonate, 9-fluorenylmethyl n-succinimidyl carbonate, 9-fluorenylmethylsuccinimidyl carbonate, 9-fluorenylmethoxycarbonyl-n-hydroxysuccinimide |
PubChem CID | 134122 |
IUPAC Name | (2,5-dioxopyrrolidin-1-yl) 9H-fluoren-9-ylmethyl carbonate |
SMILES | C1CC(=O)N(C1=O)OC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1314303
|
Thermo Scientific Chemicals
A1314303 |
1 g |
Each for $54.90
|
|
|||||
AAA1314306
|
Thermo Scientific Chemicals
A1314306 |
5 g |
Each for $137.37
|
|
|||||
AAA1314314
|
Thermo Scientific Chemicals
A1314314 |
25 g |
Each for $542.93
|
|
|||||
Description
N-(9-Fluorenylmethoxycarbonyloxy)succinimide is the most efficient reagent for the selective preparation of N-(9-fluorenylmethoxycarbonyl) derivatives of hydroxy-amino acids. Dipeptide formation is lower than with the chloroformate, Fmoc-Cl. It is also employed in the synthesis of glycopeptides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-(9-Fluorenylmethoxycarbonyloxy)succinimide is the most efficient reagent for the selective preparation of N-(9-fluorenylmethoxycarbonyl) derivatives of hydroxy-amino acids. Dipeptide formation is lower than with the chloroformate, Fmoc-Cl. It is also employed in the synthesis of glycopeptides.
Solubility
Soluble in dimethyl sulfoxide and dimethyl formamide.
Notes
Store in a cool place. Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong acids.
Chemical Identifiers
82911-69-1 | |
337.331 | |
WMSUFWLPZLCIHP-UHFFFAOYSA-N | |
134122 | |
C1CC(=O)N(C1=O)OC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24 |
C19H15NO5 | |
MFCD00010733 | |
fmoc-osu, n-9-fluorenylmethoxycarbonyloxy succinimide, fmoc-onsu, 9-fluorenylmethyl succinimidyl carbonate, fmoc n-hydroxysuccinimide ester, 9h-fluoren-9-yl methyl 2,5-dioxopyrrolidin-1-yl carbonate, 9-fluorenylmethyl n-succinimidyl carbonate, 9-fluorenylmethylsuccinimidyl carbonate, 9-fluorenylmethoxycarbonyl-n-hydroxysuccinimide | |
(2,5-dioxopyrrolidin-1-yl) 9H-fluoren-9-ylmethyl carbonate |
Specifications
82911-69-1 | |
C19H15NO5 | |
1 g | |
3569540 | |
fmoc-osu, n-9-fluorenylmethoxycarbonyloxy succinimide, fmoc-onsu, 9-fluorenylmethyl succinimidyl carbonate, fmoc n-hydroxysuccinimide ester, 9h-fluoren-9-yl methyl 2,5-dioxopyrrolidin-1-yl carbonate, 9-fluorenylmethyl n-succinimidyl carbonate, 9-fluorenylmethylsuccinimidyl carbonate, 9-fluorenylmethoxycarbonyl-n-hydroxysuccinimide | |
WMSUFWLPZLCIHP-UHFFFAOYSA-N | |
(2,5-dioxopyrrolidin-1-yl) 9H-fluoren-9-ylmethyl carbonate | |
134122 | |
98% |
150°C to 153°C | |
MFCD00010733 | |
UN3077 | |
Moisture sensitive | |
Soluble in dimethyl sulfoxide and dimethyl formamide. | |
C1CC(=O)N(C1=O)OC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24 | |
337.331 | |
337.34 | |
N-(9-Fluorenylmethoxycarbonyloxy)succinimide |
Safety and Handling
GHS H Statement
H301-H317
Toxic if swallowed.
May cause an allergic skin reaction.
P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P501c
H302-H317
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 433-520-5
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only