missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Acetoacetylmorpholine 98.0+%, TCI America™
$82.02 - $82.02
Chemical Identifiers
| CAS | 16695-54-8 |
|---|---|
| Molecular Formula | C8H13NO3 |
| Molecular Weight (g/mol) | 171.20 |
| MDL Number | MFCD00059774 |
| InChI Key | RKPILPRSNWEZJV-UHFFFAOYSA-N |
| PubChem CID | 85550 |
| IUPAC Name | 1-(morpholin-4-yl)butane-1,3-dione |
| SMILES | CC(=O)CC(=O)N1CCOCC1 |
Chemical Identifiers
| 16695-54-8 | |
| 171.20 | |
| RKPILPRSNWEZJV-UHFFFAOYSA-N | |
| 1-(morpholin-4-yl)butane-1,3-dione |
| C8H13NO3 | |
| MFCD00059774 | |
| 85550 | |
| CC(=O)CC(=O)N1CCOCC1 |
Specifications
| 16695-54-8 | |
| White | |
| C8H13NO3 | |
| 25 g | |
| CC(=O)CC(=O)N1CCOCC1 | |
| 171.20 | |
| 171.20 | |
| Crystalline Powder |
| 68°C | |
| 140°C | |
| MFCD00059774 | |
| RKPILPRSNWEZJV-UHFFFAOYSA-N | |
| 1-(morpholin-4-yl)butane-1,3-dione | |
| 85550 | |
| ≥98.0% (HPLC,N) | |
| N-Acetoacetylmorpholine |
Safety and Handling
EINECSNumber : (9)-1635
TSCA : No