missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Acetyl-D-alanine, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6345003

 View more versions of this product

Catalog No. AAH6345003


Explore available promotions

Description

Description

N-Acetyl-D-alanine may be used with other D-aminoacylated amino acids as a substrate for the identification, differentiation and characterization of D-aminoacylase(s)/amidohydrolase(s). N-acetyl-D-alanine may be used to study aglycon pocket specific binding on vancomycin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Acetyl-D-alanine may be used with other D-aminoacylated amino acids as a substrate for the identification, differentiation and characterization of D-aminoacylase(s)/amidohydrolase(s). N-acetyl-D-alanine may be used to study aglycon pocket specific binding on vancomycin.

Solubility
Slightly soluble in water.

Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
Specifications

Specifications

N-Acetyl-D-alanine
C5H9NO3
n-acetyl-d-alanine, ac-d-ala-oh, r-2-acetamidopropanoic acid, acetyl-d-alanine, n-acetylalanine, 2r-2-acetamidopropanoic acid, r-2-acetylamino-propionic acid, alanine, n-acetyl, n-acetyl-d-ala, n-acetyl d-alanine
KTHDTJVBEPMMGL-UHFFFAOYNA-N
(2R)-2-acetamidopropanoic acid
131.13
131.13
Solid
19436-52-3
MFCD00063133
Slightly soluble in water.
CC(NC(C)=O)C(O)=O
1 g
132213
98%

Chemical Identifiers

19436-52-3
131.13
KTHDTJVBEPMMGL-UHFFFAOYNA-N
132213
CC(NC(C)=O)C(O)=O
C5H9NO3
MFCD00063133
n-acetyl-d-alanine, ac-d-ala-oh, r-2-acetamidopropanoic acid, acetyl-d-alanine, n-acetylalanine, 2r-2-acetamidopropanoic acid, r-2-acetylamino-propionic acid, alanine, n-acetyl, n-acetyl-d-ala, n-acetyl d-alanine
(2R)-2-acetamidopropanoic acid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 243-066-8

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only