Learn More
N-Acetyl-D-alanine, 98%, Thermo Scientific Chemicals
$60.53 - $232.75
Chemical Identifiers
CAS | 19436-52-3 |
---|---|
Molecular Formula | C5H9NO3 |
Molecular Weight (g/mol) | 131.13 |
MDL Number | MFCD00063133 |
InChI Key | KTHDTJVBEPMMGL-UHFFFAOYNA-N |
Synonym | n-acetyl-d-alanine, ac-d-ala-oh, r-2-acetamidopropanoic acid, acetyl-d-alanine, n-acetylalanine, 2r-2-acetamidopropanoic acid, r-2-acetylamino-propionic acid, alanine, n-acetyl, n-acetyl-d-ala, n-acetyl d-alanine |
PubChem CID | 132213 |
IUPAC Name | (2R)-2-acetamidopropanoic acid |
SMILES | CC(NC(C)=O)C(O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6345003
|
Thermo Scientific Chemicals
H6345003 |
1 g |
Each for $60.53
|
|
|||||
AAH6345006
|
Thermo Scientific Chemicals
H6345006 |
5 g |
Each for $232.75
|
|
|||||
Description
N-Acetyl-D-alanine may be used with other D-aminoacylated amino acids as a substrate for the identification, differentiation and characterization of D-aminoacylase(s)/amidohydrolase(s). N-acetyl-D-alanine may be used to study aglycon pocket specific binding on vancomycin.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-Acetyl-D-alanine may be used with other D-aminoacylated amino acids as a substrate for the identification, differentiation and characterization of D-aminoacylase(s)/amidohydrolase(s). N-acetyl-D-alanine may be used to study aglycon pocket specific binding on vancomycin.
Solubility
Slightly soluble in water.
Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
Chemical Identifiers
19436-52-3 | |
131.13 | |
KTHDTJVBEPMMGL-UHFFFAOYNA-N | |
132213 | |
CC(NC(C)=O)C(O)=O |
C5H9NO3 | |
MFCD00063133 | |
n-acetyl-d-alanine, ac-d-ala-oh, r-2-acetamidopropanoic acid, acetyl-d-alanine, n-acetylalanine, 2r-2-acetamidopropanoic acid, r-2-acetylamino-propionic acid, alanine, n-acetyl, n-acetyl-d-ala, n-acetyl d-alanine | |
(2R)-2-acetamidopropanoic acid |
Specifications
19436-52-3 | |
MFCD00063133 | |
Slightly soluble in water. | |
CC(NC(C)=O)C(O)=O | |
1 g | |
132213 | |
98% | |
N-Acetyl-D-alanine |
C5H9NO3 | |
n-acetyl-d-alanine, ac-d-ala-oh, r-2-acetamidopropanoic acid, acetyl-d-alanine, n-acetylalanine, 2r-2-acetamidopropanoic acid, r-2-acetylamino-propionic acid, alanine, n-acetyl, n-acetyl-d-ala, n-acetyl d-alanine | |
KTHDTJVBEPMMGL-UHFFFAOYNA-N | |
(2R)-2-acetamidopropanoic acid | |
131.13 | |
131.13 | |
Solid |
Safety and Handling
EINECSNumber : 243-066-8
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only