missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific Chemicals N-Acetyl-D-galactosamine

$122.73 - $650.12

Chemical Identifiers

CAS 1811-31-0
Molecular Formula C8H15NO6
Molecular Weight (g/mol) 221.21
MDL Number MFCD00136044
InChI Key OVRNDRQMDRJTHS-KEWYIRBNSA-N
Synonym dsstox_cid_28637, dsstox_rid_82907, dsstox_gsid_48711, n-acetyl-, a-d-galactosamine
PubChem CID 60196345
SMILES CC(=O)N[C@H]1C(O)O[C@H](CO)[C@H](O)[C@@H]1O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ66095ME
View Documents
Thermo Scientific Chemicals
J66095ME
500 mg
Each for $122.73
 
AAJ6609503
View Documents
Thermo Scientific Chemicals
J6609503
1 g
Each for $180.62
 
AAJ6609506
View Documents
Thermo Scientific Chemicals
J6609506
5 g
Each for $650.12
 
Description

Description

N-Acetyl-D-galactosamine is the initial O-linked sugar to many serine and threonine residues in protein glycosylation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Acetyl-D-galactosamine is the initial O-linked sugar to many serine and threonine residues in protein glycosylation.

Solubility
Soluble in water at 50mg/ml

Notes
Light Sensitive & Hygroscopic. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

1811-31-0
221.21
OVRNDRQMDRJTHS-KEWYIRBNSA-N
60196345
C8H15NO6
MFCD00136044
dsstox_cid_28637, dsstox_rid_82907, dsstox_gsid_48711, n-acetyl-, a-d-galactosamine
CC(=O)N[C@H]1C(O)O[C@H](CO)[C@H](O)[C@@H]1O

Specifications

1811-31-0
White
MFCD00136044
2331340
Soluble in water at 50mg/ml
CC(=O)N[C@H]1C(O)O[C@H](CO)[C@H](O)[C@@H]1O
221.21
221.21
N-Acetyl-D-galactosamine
160°C
C8H15NO6
500 mg
dsstox_cid_28637, dsstox_rid_82907, dsstox_gsid_48711, n-acetyl-, a-d-galactosamine
OVRNDRQMDRJTHS-KEWYIRBNSA-N
N-[(2R,3S,4S,5S)-3,4,5,6-tetrahydroxy-1-oxohexan-2-yl]acetamide
60196345
Solid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 217-321-9

TSCA : No

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only