missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-acetyl-α-D-galactosamine, 99%, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0210006401

 View more versions of this product

Catalog No. ICN10006401


Explore available promotions

Description

Description

Specifications

Specifications

N-Acetyl-α-D-galactosamine
White
C8H15NO6
n-acetyl-d-galactosamine, galnac, 2-acetamido-2-deoxy-d-galactopyranose, d-galnac, 2-acetamido-2-deoxy-d-galactose, n-acetyl-d-chondrosamine, n-3r,4r,5r,6r-2,4,5-trihydroxy-6-hydroxymethyl tetrahydro-2h-pyran-3-yl acetamide, n-acetylgalactosamine, acetylgalactosamine, n-acetyl-galactosamine
CC(=O)NC1C(C(C(OC1O)CO)O)O
221.209
CHEBI:28497
Crystalline Powder
14215-68-0
0.99
1 g
OVRNDRQMDRJTHS-JAJWTYFOSA-N
N-[(2R,3R,4R,5R,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide
440552
221.2

Chemical Identifiers

14215-68-0
221.209
n-acetyl-d-galactosamine, galnac, 2-acetamido-2-deoxy-d-galactopyranose, d-galnac, 2-acetamido-2-deoxy-d-galactose, n-acetyl-d-chondrosamine, n-3r,4r,5r,6r-2,4,5-trihydroxy-6-hydroxymethyl tetrahydro-2h-pyran-3-yl acetamide, n-acetylgalactosamine, acetylgalactosamine, n-acetyl-galactosamine
CHEBI:28497
CC(=O)NC1C(C(C(OC1O)CO)O)O
C8H15NO6
OVRNDRQMDRJTHS-JAJWTYFOSA-N
440552
N-[(2R,3R,4R,5R,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide
Videos
SDS
Documents

Documents

Promotions

Promotions